Tetraoctylammonium nitrate ≥99.0%, Selectophore™ ionophore, Supelco®
Catalog # 87983-500MG
Supplier: Merck
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:528.89 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:33734-52-0
Formula:[CH₃(CH₂)₆CH₂]₄N(NO₃)
MDL Number:MFCD00525561