Tetradodecylammonium tetrakis(4-chlorophenyl)borate, Selectophore™ ionophore, Supelco®
Catalog # 87255-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:1148.32 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:100581-42-8
Formula:C₇₂H₁₁₆BCl₄N
MDL Number:MFCD00145372