Poly(vinyl chloride), carboxylated 1.8% carboxyl basis, Selectophore™ ionophore, Supelco®
Catalog # 81395-50G
Supplier: Merck
CAS Number:
Specifications
- Pk:50 g
- Seq:3368
Specifications
About this item
:
ADR:9,III
:M-Clarity™ quality level = MQ100
UN:3077
:GHS07
CAS Number:25702-80-1
Formula:(C₅H₇ClO₂)n
MDL Number:MFCD00084460