Nitrite ionophore I, Selectophore™, function tested ionophore, Supelco®
Catalog # 72590-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:1811.43 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:130549-56-3
Formula:C₁₀₂H₁₁₇ClCoN₅O₁₉
MDL Number:MFCD00213827