Order Entry
Italy
ContactUsLinkComponent
Sodium ionophore VI, Selectophore™, function tested ionophore, Supelco®
Irritant
Sodium ionophore VI, Selectophore™, function tested ionophore, Supelco®
Catalog # 71739-500MG
Supplier:  Merck
CAS Number:  
undefined
Irritant
Sodium ionophore VI, Selectophore™, function tested ionophore, Supelco®
Catalog # 71739-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :662.85 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Refrigerator
CAS Number:80403-59-4
Formula:C₃₄H₆₂O₁₂
MDL Number:MFCD00010527