
Sodium ionophore III ≥98.0% (by HPLC), Selectophore™, function tested ionophore, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:81686-22-8
:
Formula:C₃₄H₅₂N₂O₄
MDL Number:MFCD00077941
:552.79 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
Frequently Bought Together