Sodium ionophore I, Selectophore™, function tested ionophore, Supelco®
Catalog # 71732-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:641.97 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:61183-76-4
Formula:C₃₆H₇₁N₃O₆
MDL Number:MFCD00042950