Order Entry
Italy
ContactUsLinkComponent
Magnesium ionophore IV, Selectophore™, function tested ionophore, Supelco®
Magnesium ionophore IV, Selectophore™, function tested ionophore, Supelco®
Catalog # 63088-1EA
Supplier:  Merck
CAS Number:  
undefined
Magnesium ionophore IV, Selectophore™, function tested ionophore, Supelco®
Catalog # 63088-1EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Pk Info:
    50 mg in 0,5 ml

Specifications

About this item

 :
 :863.31 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:135734-39-3
Formula:C₄₉H₉₄N₆O₆
MDL Number:MFCD00144716