Order Entry
Italy
ContactUsLinkComponent
Magnesium ionophore I, Selectophore™, function tested ionophore, Supelco®
Magnesium ionophore I, Selectophore™, function tested ionophore, Supelco®
Catalog # 63082-50MG-F
Supplier:  Merck
CAS Number:  
undefined
Magnesium ionophore I, Selectophore™, function tested ionophore, Supelco®
Catalog # 63082-50MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :340.54 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:75513-72-3
Formula:C₂₀H₄₀N₂O₂
MDL Number:MFCD00043110