Order Entry
Italy
ContactUsLinkComponent
Lithium ionophore VI ≥99.0% (by HPLC), Selectophore™, function tested ionophore, Supelco®
Lithium ionophore VI ≥99.0% (by HPLC), Selectophore™, function tested ionophore, Supelco®
Catalog # 62567-100MG
Supplier:  Merck
CAS Number:  
undefined
Lithium ionophore VI ≥99.0% (by HPLC), Selectophore™, function tested ionophore, Supelco®
Catalog # 62567-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :384.51 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:106868-21-7
Formula:C₂₄H₃₂O₄
MDL Number:MFCD00037498