Potassium ionophore III, Selectophore™, function tested ionophore, Supelco®
: Merck
:99348-39-7
:
:C₄₆H₇₀N₄O₁₈
:MFCD00145150
:967.06 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Tri(2-ethylhexyl) trimellitat 1 * 100 mg
: EHRENSTORFER