Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Perfluorophenanthrene
Perfluorophenanthrene, Selectophore™ ionophore, Supelco®
Perfluorophenanthrene, Selectophore™ ionophore, Supelco®
Perfluorophenanthrene, Selectophore™ ionophore, Supelco®
56919-10ML
:
Merck
:
Perfluorophenanthrene, Selectophore™ ionophore, Supelco®
56919-10ML
:
Merck
:
Pk:
10 mL
Seq:
2275
:
:
624.116 g/mol
:
306-91-2
:
C₁₄F₂₄
:
MFCD00042596