N-Octadecanoyl nile blue, Selectophore™, function tested ionophore, Supelco®
Catalog # 24901-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:583.85 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ100
UN:2025
:GHS06
:GHS08
:GHS09
CAS Number:125829-24-5
Formula:C₃₈H₅₃N₃O₂