Order Entry
Italy
ContactUsLinkComponent
Cadmium ionophore I, Selectophore™ ionophore, Supelco®
Cadmium ionophore I, Selectophore™ ionophore, Supelco®
Catalog # 20909-50MG-F
Supplier:  Merck
CAS Number:  
undefined
Cadmium ionophore I, Selectophore™ ionophore, Supelco®
Catalog # 20909-50MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :432.73 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:73487-00-0
Formula:C₂₂H₄₄N₂O₂S₂
MDL Number:MFCD00143893