Order Entry
Italy
ContactUsLinkComponent
Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt ≥97.0%, Selectophore™ ionophore, Supelco®
Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt ≥97.0%, Selectophore™ ionophore, Supelco®
Catalog # 15180-1G
Supplier:  Merck
CAS Number:  
undefined
Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt ≥97.0%, Selectophore™ ionophore, Supelco®
Catalog # 15180-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :987.29 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:40835-97-0
Formula:C₅₆H₈₄CaO₈P₂
MDL Number:MFCD00043425