Nitrite ionophore VI, Selectophore™, function tested ionophore, Supelco®
Catalog # 03721-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:597.70 g/mol
CAS Number:228577-80-8
Formula:C₃₆H₄₆CoN₂O₂