Order Entry
Italy
ContactUsLinkComponent
 
Mesamoll®, Alkylsulphonic phenyl esters mixture, Selectophore™ ionophore, Supelco®
Catalog # 50987-5ML
Supplier:  Merck
CAS Number:  
Mesamoll®, Alkylsulphonic phenyl esters mixture, Selectophore™ ionophore, Supelco®
Catalog # 50987-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless to Light Yellow
  • Appearance (Form):
    Liquid or Viscous Liquid
  • Color (in APHA):
    ≤ 350 APHA
  • Refractive index N20/D:
    1.498 - 1.502
  • Water (Coulometric):
    ≤ 0.1 %
  • Viscosity (Rotation):
    110 - 140 mPas
  • Viscosity (Conditions):
    Neat at 20 C (DIN 45)
  • Saponification value:
    ≤ 200.0
  • RECOMMENDED RETEST PERIOD:
    36 MONTHS

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
CAS Number:91082-17-6