Order Entry
Italy
ContactUsLinkComponent
Ninhydrin reagent acc.to Stahl, yellow liquid derivatisation reagent, for TLC
Flammable
Corrosive
Irritant
Ninhydrin reagent acc.to Stahl, yellow liquid derivatisation reagent, for TLC
Catalog # 17975-100ML
Supplier:  Merck
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Ninhydrin reagent acc.to Stahl, yellow liquid derivatisation reagent, for TLC
Catalog # 17975-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :178.14 g/mol
Density:0.823 g/ml (20 °C)
ADR:3,III
 :M-Clarity™ quality level = MQ100
UN:1120
 :GHS02
 :GHS05
 :GHS07
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791