Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiourea
Thiourea ≥99.999% (metals basis), Supelco®
Thiourea ≥99.999% (metals basis), Supelco®
Thiourea ≥99.999% (metals basis), Supelco®
Catalog # 30885-5G
Supplier:
Merck
CAS Number:
Expand image
Thiourea ≥99.999% (metals basis), Supelco®
Catalog # 30885-5G
Supplier:
Merck
CAS Number:
Specifications
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
:
:
76.12 g/mol
CAS Number:
62-56-6
Formula:
NH₂CSNH₂
MDL Number:
MFCD00008067
Product Details