Order Entry
Italy
ContactUsLinkComponent
o-Cresolphthalein complexone, AnalaR NORMAPUR® Reag. Ph. Eur. indicator for metal titration
o-Cresolphthalein complexone, AnalaR NORMAPUR® Reag. Ph. Eur. indicator for metal titration
Catalog # 86166.050
Supplier:  VWR Chemicals
CAS Number:  
undefined
o-Cresolphthalein complexone, AnalaR NORMAPUR® Reag. Ph. Eur. indicator for metal titration
Catalog # 86166.050
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Suitability as indicator:
    Passes test
  • Water:
    Max. 10 %
  • Absorption Max. λ (0.1 Mol/l NaOH):
    577 --> 580 nm
  • Absorptivity (λmax;0.005 g/l 0.1 M NaOH):
    Min. 750
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

About this item

 :
 :636.62 g/mol
Melting Pt:186 °C
Storage Temperature:Ambient
EINECS:219-318-8
CAS Number:2411-89-4
Formula:C₃₂H₃₂N₂O₁₂
MDL Number:MFCD00005911