Order Entry
Italy
ContactUsLinkComponent
 
Corrosive
Nitric acid diluted (60 ml dosed with 1 ml 69% HNO₃)
Catalog # 60NIT20425
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Nitric acid diluted (60 ml dosed with 1 ml 69% HNO₃)
Catalog # 60NIT20425
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle, wide mouth
  • Pk:
    96
  • Seq:
    0096

Specifications

About this item

 :
 :63.01 g/mol
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349