Order Entry
Italy
ContactUsLinkComponent
Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane 3%Trichloroacetic Acid in Dichloromethane (w/v), BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane 3%Trichloroacetic Acid in Dichloromethane (w/v), BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR605-4X2.5L
CAS Number:  
Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane 3%Trichloroacetic Acid in Dichloromethane (w/v), BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR605-4X2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 L
  • Seq:
    5114
  • Environmentally Preferable:
  • Pk Info:
    4×2.5 L

Specifications

About this item

Synonyms: TCA
 :
 :163.39 g/mol
Flash Pt:113 Does not flash. Closed cup
EINECS:200-838-9
UN:2922
 :GHS05
 :GHS08
 :GHS07
Storage Temperature:Room Temperature
CAS Number:76-03-9
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1.33 g/cm³ (20 °C)
ADR:8 II
Merck Index:13,09700