Order Entry
Italy
ContactUsLinkComponent
 
Corrosive
Irritant
Ammonium fluoride - Hydrofluoric acid mixture, Semiconductor grade etchant solution AF 875-125 (Honeywell 17621)
Catalog # HONC40207H-2.5L
CAS Number:  
Corrosive
Irritant
Ammonium fluoride - Hydrofluoric acid mixture, Semiconductor grade etchant solution AF 875-125 (Honeywell 17621)
Catalog # HONC40207H-2.5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Ammonium Fluoride (NH₄F):
    33.8 - 35.8 %
  • Assay hydrofluoric acid (HF):
    6.2 - 6.6 %
  • Residue on ignition (SO₄):
    Max. 20 ppm
  • Arsenic (As):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.1 ppm
  • Cobalt (Co):
    Max. 0.1 ppm
  • Copper (Cu):
    Max. 0.1 ppm
  • Iron (Fe):
    Max. 1.0 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.2 ppm
  • Manganese (Mn):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.1 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • Chloride (Cl):
    Max. 4 ppm
  • Phosphate (PO₄):
    Max. 1 ppm
  • Sulfate (SO₄):
    Max. 5 ppm

Specification Test Results

About this item

CAS Number:12125-01-8
 :1341-49-7
MDL Number:MFCD03096499
Boiling Pt:100 °C
Density:1.12 g/cm³ (20 °C)
Flash Pt: Does not flash
UN:3264
ADR:8,II
 :GHS05
 :GHS07
Storage Temperature:Ambient