Order Entry
Italy
ContactUsLinkComponent
Phenol red, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Phenol red, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Supplier:  VWR Chemicals
Certificates
undefined
Phenol red, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale
Specification Test Results
  • pH Range (Yellow-Red):
    6.8 - 8.2
  • Clarity of Solution:
    Passes Test
  • Absorption Maximum λ1 (pH 1.2):
    503 - 506
  • Absorption Maximum λ2 (pH 3.0):
    430 - 435
  • Absorption Maximum λ3 (pH 6.5):
    430 - 435
  • Absorption Maximum λ4 (pH 8.8):
    557 - 560
  • Absorptivity (A1%/1 cm; λ1, on Dried Sample):
    900 - 1100
  • Absorptivity (A1%/1 cm; λ2; on Dried Sample):
    500 - 700
  • Absorptivity (A1%/1 cm; λ3, on Dried Sample):
    500 - 700
  • Absorptivity (A1%/1 cm; λ4, on Dried Sample):
    1000 - 1200
  • Insoluble in C₂H₅OH:
    Passes Test
  • Copper (Cu):
    ≤0,005
  • Iron (Fe):
    ≤0,005
  • Lead (Pb):
    ≤0,005
  • Nickel (Ni):
    ≤0,005
  • Loss on Drying (110 °C):
    ≤5