Order Entry
Italy
ContactUsLinkComponent
 
 
 
Orthophosphoric acid 85%, AR®, Macron Fine Chemicals™
  3132-46
 :  
 
 
Orthophosphoric acid 85%, AR®, Macron Fine Chemicals™
  3132-46
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Appearance:
    Clear, Colourless, Syrupy Liquid
  • Assay:
    ≥84.5%
  • Reducing substances (as H₃PO₃):
    ≤0.1%
  • Nitrate (NO₃):
    ≤0.001%
  • Chloride (Cl):
    ≤0.001%
  • Sulphate (SO₄):
    ≤0.01%
  • Calcium and Magnesium (as Ca):
    ≤0.02%
  • Arsenic (As):
    ≤0.0002%
  • Iron (Fe):
    ≤0.005%
  • Heavy Metals (as Pb):
    ≤0.002%

 

 

 :
 :98 g/mol
 :1805
 :8,II
 :GHS05
 :GHS07
 :7664-38-2
 :H₃PO₄
 :MFCD00011340