Order Entry
Italy
ContactUsLinkComponent
Diethanolamine solution 85% for synthesis (max. 0.5% ethanolamine)
Diethanolamine solution 85% for synthesis (max. 0.5% ethanolamine)
Catalog # 1.03109.5000
Supplier:  Merck
CAS Number:  
Diethanolamine solution 85% for synthesis (max. 0.5% ethanolamine)
Catalog # 1.03109.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :105.14 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:111-42-2
Formula:C₄H₁₁NO₂
MDL Number:MFCD00002843