Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Pinocembrin
Supelco® Organic Reference Standard, Pinocembrin
Catalog # P5239-50MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Pinocembrin
Catalog # P5239-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard, 95% (TLC), solid
  • CAS No.:
    480-39-7
  • Description:
    Pinocembrin
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD06858345
  • M-Clarity quality level:
    MQ100
  • Melting point:
    194 - 195 °C (lit.)
  • Molecular weight:
    256,25

Specifications

About this item