Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
O-Desmethylvenlafaxine, analytical standard, Supelco®
O-Desmethylvenlafaxine, analytical standard, Supelco®
O-Desmethylvenlafaxine, analytical standard, Supelco®
93867-50MG
:
Merck
:
O-Desmethylvenlafaxine, analytical standard, Supelco®
93867-50MG
:
Merck
:
Pk:
50 mg
Standards:
analytical standard
CAS No.:
93413-62-8
Description:
O-Desmethylvenlafaxine
Description:
O-Desmethylvenlafaxine