Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Cis/Trans F.A.M.E Column Performance Mix
Supelco® Organic Reference Standard, Cis/Trans F.A.M.E Column Performance Mix
Catalog # 40495-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Cis/Trans F.A.M.E Column Performance Mix
Catalog # 40495-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    cis/trans F.A.M.E Column Performance Mix
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H319 - H336 - H351
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Concentration:
    2,5 mg/ml in dichloromethane
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P201 - P302 + P352 - P305 + P351 + P338 - P308 + P313