Order Entry
Italy
ContactUsLinkComponent
Supelco® VETRANAL® Organic Reference Standard, Doramectin
Supelco® VETRANAL® Organic Reference Standard, Doramectin
Catalog # 33993-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® VETRANAL® Organic Reference Standard, Doramectin
Catalog # 33993-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    VETRANAL®, Analytical standard
  • CAS No.:
    117704-25-3
  • Description:
    Doramectin
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H302 - H361fd - H362 - H410
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD00894747
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)avermectin
  • Molecular weight:
    899,11
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06, GHS09
Caution: P202 - P260 - P263 - P273 - P301 + P312 - P308 + P313