Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
5-Fluorouracil, analytical standard, Supelco®
5-Fluorouracil, analytical standard, Supelco®
Catalog # 03738-100MG
Supplier:
Merck
CAS Number:
Expand image
5-Fluorouracil, analytical standard, Supelco®
Catalog # 03738-100MG
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 mg
Standards:
analytical standard
CAS No.:
51-21-8
Description:
5-Fluorouracil
Description:
5-Fluorouracil
Specifications
About this item
Product Details
Product Details