Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
(±)-Lisofylline, analytical standard, Supelco®
(±)-Lisofylline, analytical standard, Supelco®
(±)-Lisofylline, analytical standard, Supelco®
Catalog # 03716-25MG
Supplier:
Merck
CAS Number:
Expand image
(±)-Lisofylline, analytical standard, Supelco®
Catalog # 03716-25MG
Supplier:
Merck
CAS Number:
Specifications
Pk:
25 mg
Standards:
analytical standard
CAS No.:
6493-06-7
Description:
(+/-)-Lisofylline
Description:
(+/-)-Lisofylline
Specifications
About this item
Product Details
Product Details