pH PT, Proficiency Testing Material, Supelco®
Catalog # PEP010-3X100ML
Supplier: Merck
CAS Number:
Specifications
- Pk:3
- Standards:Proficiency Testing Material
- Description:pH <791> PT
Specifications
About this item
Suitable for 791 per USP. Proficiency Testing Kit containing three samples ranging from pH 5,0 to 10,00.
Certifications: This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010.