Supelco® Organic Reference Standard, Rubiadin
Catalog # 74553-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:117-02-2
- Description:Rubiadin
- Storage conditions:2...30 °C
- Hazard class description:H302 - H351
- Shipping conditions:STD
- MDL No.:MFCD02752095
- M-Clarity quality level:MQ100
- Synonym(s):1,3-Dihydroxy-2-methyl-9,10-anthracenedione; 1,3-Dihydroxy-2-methylanthraquinone
- Molecular weight:254,24
Specifications
About this item
- GHS signal: GHS07
Caution: P202 - P264 - P270 - P280 - P301 + P312 - P308 + P313