Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Pigment Red 53, analytical standard, Supelco®
Organic reference standard, Pigment Red 53, analytical standard, Supelco®
Catalog # 67195-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Pigment Red 53, analytical standard, Supelco®
Catalog # 67195-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2092-56-0
  • Description:
    Pigment Red 53, ≥98,0% (HPLC)
  • Formula:
    C₁₇H₁₂ClN₂NaO₄S
  • Description:
    Pigment Red 53
  • Form:
    Neat

Specifications

About this item