Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Kaempferol
Supelco® Organic Reference Standard, Kaempferol
Catalog # 96353-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Kaempferol
Catalog # 96353-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    520-18-3
  • Description:
    Kaempferol
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00016938
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,4′,5,7-Tetrahydroxyflavone; 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one; Robigenin
  • Molecular weight:
    286,24

Specifications

About this item

Caution: P264 - P270 - P301 + P312 - P501