Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Fumonisin B1-13C34 solution, analytical standard, Supelco®
Organic reference standard, Fumonisin B1-13C34 solution, analytical standard, Supelco®
Catalog # 33621-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Fumonisin B1-13C34 solution, analytical standard, Supelco®
Catalog # 33621-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    25 µg/ml in acetonitrile, water
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1217458-62-2
  • Description:
    Fumonisin B₁-¹³C₃₄ solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    ¹³C₃₄H₅₉NO₁₅
  • Storage temperature:
    −20°C
  • Description:
    Fumonisin B₁-¹³C₃₄ solution
  • Flash point:
    2 °C

Specifications

About this item