Supelco® Organic Reference Standard, Formononetin
Catalog # 94334-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:485-72-3
- Description:Formononetin
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00016948
- M-Clarity quality level:MQ100
- Synonym(s):7-Hydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one; 7-Hydroxy-3-(4-methoxyphenyl)chromone; 7-Hydroxy-4′-methoxyisoflavone; Biochanin B; Daidzein 4′-methyl ether; Formonetin; Formononetol
- Molecular weight:268,26