Organic reference standard, disperse red 17, analytical standard, Supelco®
Catalog # 79892-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:3179-89-3
- Description:Disperse Red 17, ≥98,0% (HPLC)
- Formula:C₁₇H₂₀N₄O₄
- Description:Disperse Red 17
- Form:Neat