Organic reference standard, Deoxynivalenol, Supelco®
Catalog # 32943-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:51481-10-8
- Description:Deoxynivalenol
- M-Clarity quality level:MQ100
- Formula:C₁₅H₂₀O₆
- Storage temperature:–20 °C
- Description:Deoxynivalenol
- Form:Neat