Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Avenanthramide A
Supelco® Organic Reference Standard, Avenanthramide A
Catalog # 30366-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Avenanthramide A
Catalog # 30366-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    108605-70-5
  • Description:
    Avenanthramide A
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD23380830
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5-Hydroxy-2-{[(2E)-3-(4-hydroxyphenyl)-1-oxo-2-propenyl]amino}benzoic acid; N-(4′-Hydroxy-(E)-cinnamoyl)-5-hydroxyanthranilic acid; Avenanthramide Bp
  • Formula:
    C₁₆H₁₃NO₅
  • Molecular weight:
    299,28
  • Flash point:
    Not applicable
  • Form:
    Neat

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501