Order Entry
Italy
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Artemisinin
Catalog # 69532-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Artemisinin
Catalog # 69532-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    63968-64-9
  • Description:
    Artemisinin
  • Storage conditions:
    RT
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00081057
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Arteannuin; Qinghaosu
  • Melting point:
    156 - 157 °C (lit.)
  • Molecular weight:
    282,33

Specifications

About this item