Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Apigenin
Supelco® Organic Reference Standard, Apigenin
Catalog # 42251-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Apigenin
Catalog # 42251-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    520-36-5
  • Description:
    Apigenin
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00006831
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4′,5,7-Trihydroxyflavone; 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone
  • Melting point:
    >300 °C (lit.)
  • Formula:
    C₁₅H₁₀O₅
  • Molecular weight:
    270,24
  • Storage temperature:
    2...8 °C
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle contents are inside inserted with fused cone.