Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Amentoflavone
Supelco® Organic Reference Standard, Amentoflavone
Catalog # 18571-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Amentoflavone
Catalog # 18571-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1617-53-4
  • Description:
    Amentoflavone
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00017470
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Didemethyl-ginkgetin; I3′,II8-Biapigenin; Tridemethylsciadopitysin
  • Formula:
    C₃₀H₁₈O₁₀
  • Molecular weight:
    538,46
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle contents are inside inserted with fused cone.