Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, α-Terpinene
Supelco® Organic Reference Standard, α-Terpinene
Catalog # 95652-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, α-Terpinene
Catalog # 95652-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    99-86-5
  • Description:
    α-Terpinene
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H226 - H302 - H304 - H317 - H319 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00001534
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-Isopropyl-4-methyl-1,3-cyclohexadiene; p-Mentha-1,3-diene
  • Molecular weight:
    136,23
  • Density:
    0,837 g/ml at 25 °C (lit.)
  • Flash point:
    50 °C - Closed cup
  • Boiling point:
    173 - 175 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS07, GHS09
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P331