Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, 3-Hydroxytyrosol
Supelco® Organic Reference Standard, 3-Hydroxytyrosol
Catalog # 91404-5MG
Nome del fornitore:  Merck
Numero CAS:  
undefined
Supelco® Organic Reference Standard, 3-Hydroxytyrosol
Catalog # 91404-5MG
Nome del fornitore:  Merck
Numero CAS:  

Specifiche

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    10597-60-1
  • Description:
    3-Hydroxytyrosol
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD01320529
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-(3,4-Dihydroxyphenyl)ethanol; 3,4-Dihydroxyphenethyl alcohol; DOPET; Homoprotocatechuyl alcohol
  • Molecular weight:
    154,16

Specifiche

Informazioni su questo articolo

  • GHS signal: GHS07
Caution: P301 + P312 + P330