Supelco® Organic Reference Standard, 17β(H),21β(H)-Hopane Solution
Catalog # 07562-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:471-62-5
- Description:17β(H),21β(H)-Hopane solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD00216315
- M-Clarity quality level:MQ100
- Synonym(s):A′-Neogammacerane
- Formula:C₃₀H₅₂
- Molecular weight:412,73
- Storage temperature:2...8 °C
- Flash point:–12 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 + P331 - P302 + P352