Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, (±)-Naringenin
Supelco® Organic Reference Standard, (±)-Naringenin
Catalog # 52186-50MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (±)-Naringenin
Catalog # 52186-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    67604-48-2
  • Description:
    (±)-Naringenin
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00006844
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one; 4′,5,7-Trihydroxyflavanone
  • Melting point:
    247 - 250 °C (lit.)
  • Molecular weight:
    272,25

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501