Order Entry
Italy
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, (±)-2,4-Dihydroxybutyric acid lithium salt
 :  Merck
 
Supelco® Organic Reference Standard, (±)-2,4-Dihydroxybutyric acid lithium salt
 :  Merck

 

  • CAS No.:
    1518-62-3
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,4-Dihydroxybutanoic acid lithium salt
  • Molecular weight:
    120,10

 

Frequently Bought Together