Supelco® Organic Reference Standard, (+)-Valencene
06808-50MG
: Merck
:
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:4630-07-3
- Description:(+)-Valencene
- Storage conditions:2...8 °C
- Hazard class description:H304
- Shipping conditions:STD
- MDL No.:MFCD00075884
- M-Clarity quality level:MQ100
- Synonym(s):(3R,4aS,5R)-4a,5-Dimethyl-3-isopropenyl-1,2,3,4,4a,5,6,7-octahydronaphthalene
- Formula:C₁₅H₂₄
- Molecular weight:204,35
- Storage temperature:2…8 °C
- Density:0,92 g/ml at 25 °C (lit.)
- Flash point:100 °C - closed cup
- Boiling point:274 °C (lit.)
: P301 + P310 - P331 - P405 - P501